Imprimer

Acide amidosulfonique 10% aqueous solution zA

Fournisseur: Bernd Kraft

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid

13649.3000 13649.4000 13649.5020
KRAF13649.3000EA 62.1 EUR
KRAF13649.3000 KRAF13649.4000 KRAF13649.5020
Acide amidosulfonique 10% aqueous solution zA
Acide amidosulfonique
Formule: H₃NO₃S
MW: 97.09 g/mol
Point d'ébullition: 247
Point de fusion: 205
Numéro MDL: MFCD00011603
Numéro CAS: 5329-14-6
EINECS: 226-218-8
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR