jetPEI® HTS DNA Transfection Reagents
Fournisseur: SARTORIUS
jetPEI® transfection reagent is a linear polyethylenimine derivative, free of components of animal origin, providing highly effective and reproducible gene delivery, perfectly suited for High Throughput Screening applications (HTS).
- Technique rapide et efficace pour la transfection d’ADN en haut débit
- Conçu pour les procédures automatisées
- Exceptionnelle reproductibilité
- Compatible avec du sérum et des antibiotiques
- Protocoles batch, reverse et forward fournis
JetPEI® possède de nombreux avantages: ce réactif est facile d’utilisation (protocoles reverse, batch et forward fournis) et est parfaitement compatible avec l’utilisation de sérum et d’antibiotiques.
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol 'New'. For duplicated products with symbol 'telephone' please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Every batch of jetPEI® is tested in-house by DNA transfection of HeLa cells. Transfection with a firefly Luciferase gene under the control of CMV promoter gives at least 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Information de livraison: une solution 150 mM de NaCl est incluse. 1 ml de réactif de transfection jetPEI® est suffisant pour effectuer 2000 transfections sur plaques 96 puits.