Imprimer
Conteneur d'urine
Fournisseur: SELECTA
1640234
216-4739EA 186 EUR
216-4739
Conteneur d'urine
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons d'urine
From plastic.
A urine sample can be taken safely and clean from the cup without opening it through the special Tapval® opening using a pipette or the supplied dispenser.
Content: 100 ml.
Attention: The product(s) on this page are outside the scope of the EU Directive 2019/904.