Imprimer
Dioxyde de zirconium ≥99% (base métaux sauf Hf) (max. 2% HfO₂)
Fournisseur: Thermo Fisher Scientific
Attention
Synonyms: Zirconium dioxide, Zirconium Oxide
11395.30 11395.A1 11395.A7
11395.30EA 102 EUR
11395.30 11395.A1 11395.A7
Dioxyde de zirconium ≥99% (base métaux sauf Hf) (max. 2% HfO₂)
Dioxyde de zirconium
Purity: 99+% CAS No.: 1314-23-4 Molecular Formula: ZrO2 Formula Weight: 123.22 Physical Form: -325 Mesh Powder Density: 5.89 Melting Point: ca 2700° Boiling Point: ca 5000° Solubility: Insoluble in water. Soluble in sulfuric acid and hydrofluoric acid. Dissolves on heating in a mixture of 2 parts H2SO4 and 1 part water Merck Reference: 14,10180 MDL No.: MFCD00011310
Unstabilized: In production of piezoelectric crystals, in high-frequency induction coils, and in ceramic glazes. Stabilized: For refractory furnace linings, and in crucibles
Formule: ZrO₂
MW: 123.22 g/mol
Point d'ébullition: ∼5000 °C
Point de fusion: ∼2700 °C
Densité: 5.89
Température de stockage: Température ambiante
Numéro MDL: MFCD00011310
Numéro CAS: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180