Imprimer
Acide sulfurique ≥95% ≥95%, TraceSELECT™ Ultra pour analyse des ultratraces, Fluka™
Fournisseur: Honeywell Chemicals
Danger
77239-250ML 77239-1L 77239-500ML
77239-250MLEA 436 EUR
77239-250ML 77239-1L 77239-500ML
Acide sulfurique ≥95% ≥95%, TraceSELECT™ Ultra pour analyse des ultratraces, Fluka™
Acide sulfurique
TraceSELECT™ Ultra and TraceSELECT
  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
Formule: H₂SO₄
MW: 98.08 g/mol
Point d'ébullition: 310 °C (1.013 hPa)
Point de fusion: –14…–10 °C
Densité: 1.840 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00064589
Numéro CAS: 7664-93-9
EINECS: 231-639-5
ONU: 1830
ADR: 8 II
Commander maintenant
Résultats des tests de spécification
Assay 94.5 - 97.0 %
Silver (Ag) Max. 0.01 ppb
Aluminium (Al) Max. 0.5 ppb
Arsenic (As) Max. 1 ppb
Barium (Ba) Max. 0.02 ppb
Beryllium (Be) Max. 0.01 ppb
Bismuth (Bi) Max. 0.01 ppb
Calcium (Ca) Max. 2 ppb
Cadmium (Cd) Max. 0.01 ppb
Cerium (Ce) Max. 0.01 ppb
Cobalt (Co) Max. 0.01 ppb
Chromium (Cr) Max. 0.05 ppb
Cesium (Cs) Max. 0.01 ppb
Copper (Cu) Max. 0.5 ppb
Dysprosium (Dy) Max. 0.01 ppb
Erbium (Er) Max. 0.01 ppb
Europium (Eu) Max. 0.01 ppb
Iron (Fe) Max. 0.5 ppb
Gallium (Ga) Max. 0.01 ppb
Gadolinium (Gd) Max. 0.01 ppb
Hafnium (Hf) Max. 0.01 ppb
Mercury (Hg) Max. 0.5 ppb
Holmium (Ho) Max. 0.01 ppb
Indium (In) Max. 0.01 ppb
Potassium (K) Max. 0.5 ppb
Lanthanum (La) Max. 0.01 ppb
Lithium (Li) Max. 0.02 ppb
Lutetium (Lu) Max. 0.01 ppb
Magnesium (Mg) Max. 0.2 ppb
Manganese (Mn) Max. 0.02 ppb
Molybdenum (Mo) Max. 0.05 ppb
Sodium (Na) Max. 1 ppb
Niobium (nb) Max. 0.01 ppb
Neodymium (nd) Max. 0.01 ppb
Nickel (Ni) Max. 0.1 ppb
Lead (Pb) Max. 0.05 ppb
Palladium (Pd) Max. 0.01 ppb
Praseodymium (Pr) Max. 0.01 ppb
Platinum (Pt) Max. 0.01 ppb
Rubidium (Rb) Max. 0.01 ppb
Rhodium (Rh) Max. 0.01 ppb
Antimony (Sb) Max. 0.05 ppb
Scandium (Sc) Max. 0.02 ppb
Selenium (Se) Max. 0.001 ppm
Samarium (Sm) Max. 0.01 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.01 ppb
Terbium (Tb) Max. 0.01 ppb
Tellurium (Te) Max. 0.01 ppb
Thorium (Th) Max. 0.01 ppb
Titanium (Ti) Max. 0.1 ppb
Thallium (Tl) Max. 0.01 ppb
Thulium (Tm) Max. 0.01 ppb
Uran (U) Max. 0.01 ppb
Vanadium (V) Max. 0.01 ppb
Tungsten (W) Max. 0.01 ppb
Yttrium (Y) Max. 0.01 ppb
Ytterbium (Yb) Max. 0.01 ppb
Zinc (Zn) Max. 0.5 ppb
Zirconium (Zr) Max. 0.02 ppb
Vapor Pressure 1 mmHg ( 146 °C)
Vapor Density <0.3 (25 °C, vs air)