Imprimer
Lithium aluminium hydrure ≥95%
Fournisseur: Thermo Fisher Scientific
Danger
13442.14 13442.22 13442.36 13442.51
13442.14EA 58.7 EUR
13442.14 13442.22 13442.36 13442.51
Lithium aluminium hydrure ≥95%
Lithium aluminium hydrure
Purity: 95% min
CAS No.: 16853-85-3
Molecular Formula: LiAlH4
Formula Weight: 37.95
Physical Form: Powder
Density: 0.917
Appearance: White to grey
Odour: Odourless
Melting Point: >125° dec.
Solubility: Decomposes in water. Soluble in ether
Merck Reference: 14,349
UN No.: 1410
MDL No.: MFCD00011075
Reducing agent for pharmaceuticals, perfumes, and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizations
Formule: H₄AlLi
Point de fusion: >125 °C (déc.)
Densité: 0,917
Température de stockage: Température ambiante
Numéro MDL: MFCD00011075
Numéro CAS: 16853-85-3
EINECS: 240-877-9
ONU: 1410
ADR: 4.3,I
Merck Index: 14,349