5-Fluoro-2-nitrotoluène ≥98%
Fournisseur: Thermo Fisher Scientific
Purity: 98+%
CAS No.: 446-33-3
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Flash Point: 87°(188°F)
Refractive Index: 1.5280
Melting Point: ca 20°
Boiling Point: 97-98°/10mm
Beilstein Reference: 2046652
UN No.: 2810
MDL No.: MFCD00007283