Imprimer

ALFAA16545

Fournisseur: Thermo Scientific
A16545.18EA 290 EUR
ALFAA16545.18
ALFAA16545
Purity: 95%
CAS No.: 503-60-6
Molecular Formula: C5H9Cl
Formula Weight: 104.57
Storage Temperature: 2°C to 8°C
Flash Point: 13°(56°F)
Refractive Index: 1.4490
Boiling Point: 107-109°
UN No.: 1993
MDL No.: MFCD00000982
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR