Imprimer
Benzaldéhyde ≥98%
Fournisseur: Thermo Fisher Scientific
Attention
30749.A1 30749.A5 30749.30
30749.A1EA 39.9 EUR
30749.A1 30749.A5 30749.30
Benzaldéhyde ≥98%
Benzaldéhyde
Purity: 98%
CAS No.: 100-52-7
Molecular Formula: C7H6O
Formula Weight: 106.12
Flash Point: 62°(143°F)
Physical Form: Liquid
Density: 1.047
Refractive Index: 1.5450
Sensitivity: Air Sensitive
Appearance: Colourless
Odour: Almond-like
Melting Point: -26°
Boiling Point: 178-179°
Beilstein Reference: 471223
Merck Reference: 14,1058
UN No.: 1990
MDL No.: MFCD00003299
Formule: C₆H₅CHO
Point d'ébullition: 178…179 °C
Point de fusion: –26 °C
Densité: 1,047
Point d'éclair: 62 °C (143 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00003299
Numéro CAS: 100-52-7
EINECS: 202-860-4
ONU: 1990
ADR: 9,III
Merck Index: 14,1058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98811dd8cad9340872b5c4b5bfb305860eb1f81a&vid=934c1b87225e77b213e66c334d8f27ef4d924847&prid=web&clid=SAPDAM