Imprimer
Oxobis(2,4-pentanedionato-O,O')vanadium (IV)
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)
81119.18 81119.09 81119.30
81119.18EA 91.8 EUR
81119.18 81119.09 81119.30
Oxobis(2,4-pentanedionato-O,O')vanadium (IV)
Oxobis(2,4-pentanedionato-O,O')vanadium (IV)
CAS No.: 3153-26-2 Molecular Formula: C10H14O5V Formula Weight: 265.16 Physical Form: Powder Density: 1.4 Melting Point: 256-259° Boiling Point: 140°/13mm Solubility: Moderately soluble in acetone, ether and chloroform. Soluble in ethanol and benzene UN No.: 3285 MDL No.: MFCD00000032
As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment
Formule: C₁₀H₁₄O₅V
MW: 265.16 g/mol
Point d'ébullition: 140 °C (13 mmHg)
Point de fusion: 256…259 °C
Densité: 1.4
Température de stockage: Température ambiante
Numéro MDL: MFCD00000032
Numéro CAS: 3153-26-2
EINECS: 221-590-8
ONU: 3285
ADR: 6.1 III