Imprimer
Béchers, forme haute, Verre borosilicaté DURAN®
Fournisseur: Lenz Laborglas GmbH & CO.KG
07004028 07004037 07004043 07004049 07004055 07004061 07004067 07004070 07004073 07004076
LENZ07004028EA 96.9 EUR
LENZ07004028 LENZ07004037 LENZ07004043 LENZ07004049 LENZ07004055 LENZ07004061 LENZ07004067 LENZ07004070 LENZ07004073 LENZ07004076
Béchers, forme haute, Verre borosilicaté DURAN®
Béchers
DURAN® borosilicate glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.