Imprimer
Propanol-1, anhydrous 99,9
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
41465.AK 41465.K2 41465.K7
41465.AKEA 47.5 EUR
41465.AK 41465.K2 41465.K7
Propanol-1, anhydrous 99,9
Propanol-1
Purity: 99.9%
CAS No.: 71-23-8
Molecular Formula: CH3CH2CH2OH
Formula Weight: 60.10
Flash Point: 15°(59°F)
Physical Form: Liquid
Density: 0.804
Refractive Index: 1.3850
Odour: Alcohol-like
Melting Point: -127°
Boiling Point: 97°
Beilstein Reference: 1098242
Merck Reference: 14,7842
UN No.: 1274
MDL No.: MFCD00002941
Solvent
Formule: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Point d'ébullition: 97 °C
Point de fusion: –127 °C
Densité: 0.804
Point d'éclair: 15 °C (59 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00002941
Numéro CAS: 71-23-8
EINECS: 200-746-9
ONU: 1274
ADR: 3 II
Merck Index: 14,7842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM