Imprimer
Butanol-1 99% HPLC
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
22925.K2 22925.M1
22925.K2EA 80.9 EUR
22925.K2 22925.M1
Butanol-1 99% HPLC
Butanol-1
Purity: 99%
CAS No.: 71-36-3
Molecular Formula: CH3(CH2)3OH
Formula Weight: 74.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 0.810
Refractive Index: 1.3990
Odour: Alcohol-like
pH: 7
Melting Point: -90 to -89°
Boiling Point: 117-118°
Beilstein Reference: 969148
Merck Reference: 14,1540
UN No.: 1120
MDL No.: MFCD00002964
Formule: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Point d'ébullition: 117…118 °C
Point de fusion: –90…–89 °C
Densité: 0.810
Point d'éclair: 35 °C (95 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00002964
Numéro CAS: 71-36-3
EINECS: 200-751-6
ONU: 1120
ADR: 3 III
ATTEINDRE: 01-2119484630-38
Merck Index: 14,1540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM