Imprimer
Cérium trinitrate hexahydraté ≥99.5% (REO, base d'oxyde de terre rare), REacton®
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate
11329.09 11329.22 11329.36 11329.A1
11329.09EA 52.9 EUR
11329.09 11329.22 11329.36 11329.A1
Cérium trinitrate hexahydraté ≥99.5% (REO, base d'oxyde de terre rare), REacton®
Cérium trinitrate hexahydraté
Purity: 99.5% CAS No.: 10294-41-4 Molecular Formula: CeN3O9.6H2O Formula Weight: 434.23 (326.13anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic pH: 3 Solubility: Soluble in water, alcohol, acetone Merck Reference: 14,2000 UN No.: 1477 MDL No.: MFCD00149631
Separation of cerium from other rare earths, catalyst for hydrolysis of phosporic acid esters
Formule: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Point d'ébullition: 200
Point de fusion: 39
Densité: 2.377 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149631
Numéro CAS: 10294-41-4
EINECS: 233-297-2
ONU: 1477
ADR: 5.1 III
Merck Index: 14,2000