Imprimer
Tétrahydrofuranne 99 Stabilised
Fournisseur: Thermo Fisher Scientific
Danger
Synonyms: Butylene oxide, THF, Diethylene oxide
Stabilization: 250 ppm BHT
L13304.AP L13304.AU L13304.0F
L13304.APEA 47.9 EUR
L13304.AP L13304.AU L13304.0F
Tétrahydrofuranne 99 Stabilised
Tétrahydrofuranne
Stabilized with 250-350 ppm BHT
Purity: 99% CAS No.: 109-99-9 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -17°(1°F) Refractive Index: 1.4070 Odour: Ether-like Melting Point: -108° Boiling Point: 66° Beilstein Reference: 102391 Merck Reference: 14,9211 UN No.: 2056 MDL No.: MFCD00005356
Formule: C₄H₈O
MW: 72.11 g/mol
Point d'ébullition: 66 °C
Point de fusion: –108 °C
Densité: 0.889
Point d'éclair: –17 °C (1 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00005356
Numéro CAS: 109-99-9
EINECS: 203-726-8
ONU: 2056
ADR: 3 II
ATTEINDRE: 05-2114615188-48
Merck Index: 14,9211
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM