Imprimer
Tungstate de disodium dihydraté 99.0-101.0% ACS
Fournisseur: Thermo Fisher Scientific
Attention
Synonyms: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate
36489.22 36489.36
36489.22EA 79.4 EUR
36489.22 36489.36
Tungstate de disodium dihydraté 99.0-101.0% ACS
Tungstate de disodium dihydraté
Purity: 99.0-101.0% CAS No.: 10213-10-2 Molecular Formula: Na2WO4.2H2O Formula Weight: 329.86 (293.83anhy) Physical Form: Powder Density: 3.250 Appearance: White Odour: Odourless Melting Point: 100° -2H2O Merck Reference: 14,8698 MDL No.: MFCD00149190
Formule: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Point de fusion: 100 °C (-2H₂O)
Densité: 3.250
Température de stockage: Température ambiante
Numéro MDL: MFCD00149190
Numéro CAS: 10213-10-2
EINECS: 236-743-4
Merck Index: 14,8698