Imprimer
Préfiltres en microfibres de verre
Fournisseur: Corning
431410 431411 431412 431413
734-2297EA 83.12 EUR
734-2297 734-2298 734-2299 734-2300
Préfiltres en microfibres de verre
Filtres Filtres de fibres de verre
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.