Imprimer
CAPS (Acide 3-(cyclohexylamino)propanesulfonique) ≥99%, Haute Pureté
Fournisseur: VWR Chemicals
Synonyms: 3-Cyclohexylaminopropanesulphonic acid
0365-1KGEA 549 EUR
0365-1KG 0365-250G 0365-500G
CAPS (Acide 3-(cyclohexylamino)propanesulfonique) ≥99%, Haute Pureté
CAPS (Acide 3-(cyclohexylamino)propanesulfonique)
Biological buffers are useful for cell culture invitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological.
Formule: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
Point de fusion: 324 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00003837
Numéro CAS: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773
Commander maintenant
Résultats des tests de spécification
pKa @25 °C 10.3 - 10.5
Purity ≤99%