Imprimer
Microscopes polarisants, BA310-POL
Fournisseur: MOTIC
1100100402242 1101001703961 1101001703941 1101001703951 1101001703981 1101001703971
630-1822EA 4660 EUR
630-1822 MOTI1101001703961 MOTI1101001703941 MOTI1101001703951 MOTI1101001703981 MOTI1101001703971
Microscopes polarisants, BA310-POL
Microscopes
These models offer higher optical performance and modularity.
  • Polariser mounted under the condenser holder rotates 360° with click stop at 90°
  • Highly stable, robust symmetrical pyramid design and T-shaped base
  • Anti-fungal chemical treatment to inhibit germs growth
  • Plan achromatic, stain-free objectives
  • One eyepiece features a crosshair reticle
Suitable for life science, geology, material science and forensics.
Information de livraison: Supplied with an Allen key, a blue filter and a dust cover.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Tête Inclinaison de 30°
Oculaires Grand champ 10×/20 mm avec pointeur
Tourelle 4 positions inversées
Objectifs Achromatiques 4×, 10×, 40×, 60×
Condenseur Abbe N.A. 0,90/1,25 with iris diaphragm
Mise au point Coaxiale micro et macrométrique
Éclairage Halogen