Imprimer
β-D-Glucopyrannoside d'α-hydroxy-o-tolyle, en poudre
Fournisseur: MP Biomedicals
Synonyms: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin
0210283305 0210283380 0210283310 0210283325
ICNA0210283305EA 61.7 EUR
ICNA0210283305 ICNA0210283380 ICNA0210283310 ICNA0210283325
β-D-Glucopyrannoside d'α-hydroxy-o-tolyle, en poudre
β-D-Glucopyrannoside d'α-hydroxy-o-tolyle
Salicin is an alcoholic β-glucoside derived from willow bark that produces anti-inflammatory (including analgesic and antipyretic) effects very similar to that of aspirin Salicin is a substrate for β-glucosidase.
Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.
Biochem/physiol Actions: It acts as a non-selective COX inhibitor, exhibiting IC50 values > 100 µM for COX-1 and COX-2.
Freely soluble in water (50 mg/ml-clear,colorless solution); soluble in alkalies, pyridine, glacial acetic acid; practically insoluble in ether and chloroform.
Formule: C₁₃H₁₈O₇
Point de fusion: 199 - 201 °C
Densité: 1.42 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00006590
Numéro CAS: 138-52-3
EINECS: 205-331-6
Merck Index: 12,08476
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
FTIR Conforms to Standard
194 - 204 °C Melting Point
Optical Rotation −65° ± 5° (c=3, water)