Imprimer
Wipe dispenser, wall mountable, zetMatic®
Fournisseur: ZVG
zetMatic®
115-2741EA 98.1 EUR
115-2741
Wipe dispenser, wall mountable, zetMatic®
Distributeurs de lingettes
Wall-mountable dispenser made of steel with an aluminum powder coating. Suitable for rolls with a width of max. 42 cm.
  • With waste bag holder
  • Wipe rolls can be easily exchanged
  • For wall mounting