Imprimer

Anti-MCM2 Mouse Monoclonal Antibody [clone: ABT-MCM2]

Fournisseur: ANTIBODIES.COM
A100019-100 A100019-50
ANTIA100019-100EA 475 EUR
ANTIA100019-100 ANTIA100019-50
Anti-MCM2 Mouse Monoclonal Antibody [clone: ABT-MCM2]
Anticorps
Mouse monoclonal (ABT-MCM2) antibody to MCM2 for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: MCM2
Clonality: Monoclonal
Clone: ABT-MCM2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR