Imprimer

Anti-ALDH1L1 Rabbit Polyclonal Antibody

Fournisseur: Antibodies.com
A85310-100 A85310-50
ANTIA85310-100EA 520 EUR
ANTIA85310-100 ANTIA85310-50
Anti-ALDH1L1 Rabbit Polyclonal Antibody
Anticorps
Rabbit polyclonal antibody to ALDH1L1 for WB and ICC/IF with samples derived from Human, Rat and Mouse.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:2000 to 1:5000, ICC/IF: 1:100

Type: Primary
Antigen: ALDH1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR