Imprimer

Anti-Fibrinogen Rat Monoclonal Antibody [clone: KT10]

Fournisseur: ANTIBODIES.COM
A83-100 A83-200
ANTIA83-100EA 445 EUR
ANTIA83-100 ANTIA83-200
Anti-Fibrinogen Rat Monoclonal Antibody [clone: KT10]
Anticorps
Rat monoclonal (KT10) antibody to Fibrinogen for ELISA with samples derived from human.

  • Validated applications: ELISA
  • Recommended dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: Fibrinogen
Clonality: Monoclonal
Clone: KT10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR