Imprimer

Anti-S100 alpha Mouse Monoclonal Antibody [clone: ABT-S100A1]

Fournisseur: ANTIBODIES.COM
A98939-100 A98939-50
ANTIA98939-100EA 475 EUR
ANTIA98939-100 ANTIA98939-50
Anti-S100 alpha Mouse Monoclonal Antibody [clone: ABT-S100A1]
Anticorps
Mouse monoclonal (ABT-S100A1) antibody to S100A1 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: S100 alpha
Clonality: Monoclonal
Clone: ABT-S100A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR