Imprimer

Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [Clone: 1D5]

Fournisseur: ANTIBODIES.COM
A254278-100 A254278-500
ANTIA254278-100EA 355 EUR
ANTIA254278-100 ANTIA254278-500
Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [Clone: 1D5]
Anticorps
Mouse monoclonal [1D5] antibody to Estrogen Receptor alpha for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1:100 to 1:200

Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR