Imprimer
Twister® Automatic Film Coverslipper
Fournisseur: MEDITE
03-5000-01 03-5010-00
MEDT03-5000-01EA 32430 EUR
MEDT03-5000-01 MEDT03-5010-00
Twister® Automatic Film Coverslipper
Colleuses de lamelles
The Twister® is an automated film coverslipper.
The Twister® permits the use of slide staining holders with a 20 or 30 piece capacity. A total of 1200 preparations can be covered per hour. Empty holders, and holders loaded with finished slides are automatically detected and passed down in the output.
The required amount of Xylene can be individually regulated by an adjusting screw. Furthermore, a bottle ensures that excess Xylene can be safely collected and disposed of. The Twister® has an integrated charcoal filter, which prevents the emission of toxic fumes to protect the user. The unit may also be connected to an external exhaust.
The glass panel front of the device enables continuous and full insight into interior processes, as well as easy access for the replacement of coverslipping film. The front panel otherwise protects against all environmental influences. A cutout in the door allows easy access to the loading station. A lucid colour touch screen permits quick and easy setting of processing parameters.
Informations de commande: Accessories not included.