Imprimer
Paraplast Plus® pour tissu, pour enrobage
Fournisseur: SIGMA-ALDRICH MICROSCOPY
M-Clarity™ (Niveau de qualité) = MQ100
P3683-1KGEA 55.6 EUR
P3683-1KG
Paraplast Plus® pour tissu, pour enrobage
Paraffine en mélange avec polymères synthétiques
Paraplast Plus® is a tissue embedding medium comprising highly purified paraffin and polyisobutylene with approximately 0.8% DMSO added to improve infiltration, sectioning, and penetration into tissues. It is recommended for tissues that are difficult to process with regular Paraplast®. Paraplast Plus® cuts continuous ribbons at 2 μm thickness or short ribbons at 1 μm thickness.
  • Paraplast Plus® has been used in the examination of vertebral specimens of Malacoraja senta
  • It has also been used in the identification and expression analysis of PIN genes in rice
  • It has been used in the study of glucose transport mechanisms in uterine tissues during pregnancy
  • It has been used to embed Bouin's fixed carp tissues, rat ventral prostatic fragments, and common bean (Phaseolus vulgaris L.) nodules
Point de fusion: 56,..57 °C (lit.)                      
Température de stockage: Room Temperature
Numéro CAS: 8002-74-2