Imprimer
Clamp Containers with Silicone Seal
Fournisseur: Sampling Systems
A566-05 A566-10 A566-20 A566-25 A566-30
SAMPA566-05EA 1050 EUR
SAMPA566-05 SAMPA566-10 SAMPA566-20 SAMPA566-25 SAMPA566-30
Clamp Containers with Silicone Seal
Récipients de collecte d'échantillon Conteneurs à usage universel
Clamp containers are ideal for use in the pharmaceutical industry and other hygiene critical industries.
  • High-quality and heavy-duty containers with a tri-ferrule lid
  • Available in a wide range of sizes
  • Supplied with side handles and silicone seal
  • Available from stock
  • Fast delivery
This range of high-quality and heavy-duty clamp containers are ideal for use across hygiene critical industries. The robust design includes side handles to ensure safe and easy transport and storage. The mirror polish finish ensure easy cleaning after use.