Imprimer

Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 647)

Fournisseur: AAT BIOQUEST
100510F0 100510F1
AATB100510F0EA 371 EUR
AATB100510F0 AATB100510F1
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 647)
Anticorps
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR