Imprimer

Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/iFluor® 750)

Fournisseur: AAT BIOQUEST
103801R0 103801R1 103801R2
AATB103801R0EA 319 EUR
AATB103801R0 AATB103801R1 AATB103801R2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE/iFluor® 750)
Anticorps
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE/iFluor® 750
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR