Imprimer

Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 790)

Fournisseur: AAT BIOQUEST
104410M0 104410M1
AATB104410M0EA 371 EUR
AATB104410M0 AATB104410M1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 790)
Anticorps
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 790
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR