Imprimer

Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 860)

Fournisseur: AAT BIOQUEST
100330R0 100330R1
AATB100330R0EA 371 EUR
AATB100330R0 AATB100330R1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 860)
Anticorps
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR