Imprimer

Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 647)

Fournisseur: AAT BIOQUEST
102800F0 102800F1
AATB102800F0EA 371 EUR
AATB102800F0 AATB102800F1
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 647)
Anticorps
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR