Imprimer

Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 647)

Fournisseur: AAT BIOQUEST
116100F0 116100F1
AATB116100F0EA 371 EUR
AATB116100F0 AATB116100F1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 647)
Anticorps
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR