Imprimer

Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]

Fournisseur: AAT Bioquest
AATB10041000EA 249 EUR
AATB10041000
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]
Anticorps
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Attention: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR