Imprimer
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® A7)
Fournisseur: AAT BIOQUEST
100320S0 100320S1
AATB100320S0EA 371 EUR
AATB100320S0 AATB100320S1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® A7)
Anticorps
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Attention: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human