Imprimer
12-Methoxydodecanoic acid ≥98% (par CCM)
Fournisseur: ENZO LIFE SCIENCES
BMLG2340010 BMLG2340050
ENZOBMLG2340010EA 85.8 EUR
ENZOBMLG2340010 ENZOBMLG2340050
12-Methoxydodecanoic acid ≥98% (par CCM)
12-Methoxydodecanoic acid
A myristate analog which has similar chain length but reduced hydrophobicity compared to myristate. Inhibits replication of HIV-1. Its specific mechanism of action is not clear but it is believed to either inhibit the activity of N-myristoyltransferase or modulate the function of target acylated proteins.
  • Myristate analog
White to off-white solid.
Soluble in ethanol (25 mg/mL) and DMSO (25 mg/mL).
Formule: C₁₃H₂₆O₃
Numéro MDL: MFCD00899847
Numéro CAS: 92169-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65c59413cc056d8ca9f07c472391d2a6a623e6f8&vid=0c76b60f74f4ae1e83888ec462f877140ac20031&prid=web&clid=SAPDAM