Imprimer
Visual colour kits for water testing, VISOCOLOR®
Fournisseur: MACHEREY-NAGEL
931098 931298
MANA931098EA 65.8 EUR
MANA931098 MANA931298
Visual colour kits for water testing, VISOCOLOR®
Kits de tests environnementaux
Colorimetric determination of zinc using zincon method.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
Zinc is one of the most often used metals for surface finishing. Its content in waste water from e. g. electroplating companies has to be monitored regularly.
Suitable for sea water testing after dilution.