Imprimer

Ammoniaque 1 mol/l (1 N)

Fournisseur: Bernd Kraft
Attention

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

03275.3000 03275.4000 03275.5020
KRAF03275.3000EA 61.5 EUR
KRAF03275.3000 KRAF03275.4000 KRAF03275.5020
Ammoniaque 1 mol/l (1 N)
Ammoniaque
Formule: NH₃
MW: 35.05 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00066650
Numéro CAS: 1336-21-6
EINECS: 215-647-6
ONU: 2672
ADR: 8 III

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR