Imprimer

tri-Potassium citrate

Fournisseur: LOHMANN

16237.9025 16237.9050 162372.9050
LOHM16237.9025EA 0 EUR
LOHM16237.9025 LOHM16237.9050 LOHM162372.9050
tri-Potassium citrate
tri-Potassium citrate
Formule: C₆H₅K₃O₇
Numéro CAS: 866-84-2
EINECS: 212-755-5
Merck Index: 12,07785

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR