Imprimer
ESD Safety Shoes, Low, LUANA, S1
Fournisseur: ELTEN
749125/37 749125/35 749125/36 749125/38 749125/39 749125/40 749125/41 749125/42
ELTN749125/37EA 96.9 EUR
ELTN749125/37 ELTN749125/35 ELTN749125/36 ELTN749125/38 ELTN749125/39 ELTN749125/40 ELTN749125/41 ELTN749125/42
ESD Safety Shoes, Low, LUANA, S1
Chaussures et bottes
Mesh material with recycled material content/Microfibre with recycled material content.
  • Breathable lining with recycled material content
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD grey with recycled material content
  • Soft fleece ESD insole
  • TPU/PU sole MAXXIMO
  • Steel toe cap
Designed for use in dry industry, storage, transport, and assembly work areas where there is a risk of electrostatic discharge (ESDS/ESD). Shoes are also suitable for use in workplaces on hard undergrounds. The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, creating more energy in every step.
Certifications: EN ISO 20345 S1 FO/SR, form A.