Imprimer
Manganèse (IV) oxyde tech. 90%, grade Technique, activé
Fournisseur: Thermo Fisher Scientific
Attention
Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
L13253.22 L13253.09 L13253.36
L13253.22EA 34.9 EUR
L13253.22 L13253.09 L13253.36
Manganèse (IV) oxyde tech. 90%, grade Technique, activé
Manganèse (IV) oxyde
Manganese(IV) oxide is suitable for use in batteries. It is widely used for the selective oxidation of allylic and benzylic alcohols to aldehydes or ketones, as a co-oxidant in combination with 1,4-benzoquinone and in the allylic oxidation of olefins catalyzed by Palladium(II)­ acetate. It acts a reagent for oxidations. It is the source of manganese and all its compounds, largely used in manufacturing of manganese steel, for making amethyst glass, decolorizing glass, painting on porcelain, faience and majolica. The precipitate is used in electrotechnics, pigments, browning gun barrels, drier for paints and varnishes, printing and dyeing textiles.
Formule: MnO₂
MW: 86.94 g/mol
Point de fusion: ∼ 535 °C (déc.)
Densité: 5.026
Température de stockage: Température ambiante
Numéro MDL: MFCD00003463
Numéro CAS: 1313-13-9
Merck Index: 14,5730