Imprimer
di-Sodium succinate hexahydraté
Fournisseur: Thermo Fisher Scientific
Attention
Synonyms: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate
33386.30 33386.A1
33386.30EA 47 EUR
33386.30 33386.A1
di-Sodium succinate hexahydraté
Succinate de disodium hexahydraté
CAS No.: 6106-21-4 Molecular Formula: C4H4Na2O4.6H2O Formula Weight: 270.14 (162.05anhy) Physical Form: Granular/crystalline powder Sensitivity: Hygroscopic Appearance: White Melting Point: 120° -6H2O Beilstein Reference: 3921044 Merck Reference: 14,8679 MDL No.: MFCD00149117
Formule: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Point de fusion: 120 °C (-6H₂O)
Densité: 1.529 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149117
Numéro CAS: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754