Imprimer

Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 647)

Fournisseur: AAT BIOQUEST
16642 16809
AATB16642EA 120 EUR
AATB16642 AATB16809
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 647)
Anticorps
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Attention: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR