Imprimer
Streptavidin, RPE-Cy7
Fournisseur: AAT BIOQUEST
16916 16917
AATB16916EA 143 EUR
AATB16916 AATB16917
Streptavidin, RPE-Cy7
Protéines et Peptides
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Extremely high biotin-binding affinity
  • High quality conjugate
Attention: For research use only.