Screen Quest™ No Wash Potassium Channel Assay Kit
Fournisseur: AAT Bioquest
Potassium (K⁺) ion channel plays an important role in regulating fundamental biological processes including heart rate, hormone and neurotransmitter secretion, water and electrolyte balance.
- Cell permeable potassium probe
- No wash step required
- Robust and adaptable protocol
- Multiple research applications
Attention: For research use only.