Imprimer
Hygostar PB 4B + PB 6B, PP, PE Protective Gowns Fully Laminated
Fournisseur: MENSCH FRANZ
277261 277264
MENS277261EA 211 EUR
MENS277261 MENS277264
Hygostar PB 4B + PB 6B, PP, PE Protective Gowns Fully Laminated
Vêtements à usage unique
The protective gowns type PB 4B + PB 6B is made of polypropylene (PP) and CAT3 certificated. Therefore it protects against irreversible or fatal risks in the pharmaceutical or medical sector. Its' arms, front and back are fully coated with PE to keep the gown very tight against liquids and splashes. Furthermore there are comfortable knitted cuffs on the sleeves for an individual fit.
  • Made of virus-proof material with comfortable knitted cuffs
  • Barrier against infectious and biological hazards (EN14126)
  • Continuous PE lamination on arms, front and back
  • High tightness
  • Protection against irreversible or fatal risks
The protective gowns type PB 4B + PB 6B can be used in the field of pharmacy and medicine as well as in medical emergency services.
Type of inner packaging: PE
Type of outer packaging: Cardboard
Material: PP with PE coating
Certifications: CE 0624 PSA VO (EU) 2016-425, CAT 3, EN 14126.