Imprimer
Aggressive Adhesive Multi-Purpose Clear Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Fournisseur: Brady
170873 170874
BRDY170873EA 63.85 EUR
BRDY170873 BRDY170874
Aggressive Adhesive Multi-Purpose Clear Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Étiquettes
Label panels and equipment with the B-432 clear polyester label material, allowing the applied surface colors to transmit around the printed label text. Print almost anywhere with a Brady handheld label maker.
  • Versatile - Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Tough - Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Adhesive - Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Application: Asset tracking, barcode labeling, component and equipment labeling, data and telecommunications labeling, electrical labeling, general identification, inventory and inspection labeling, warehouse marking
Gloss finish, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant B-432 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: UL recognized to UL969, CSA accepted.