TAAG F14 SPID
Fournisseur: KREGLINGER EUROPE
Ready-to-use multiplex real-time PCR kit for the simultaneous detection and identification of Listeria monocytogenes from enriched samples using TAAG E41 pathogens food or TAAG E31 pathogens surfaces.
Attention: Kit components are stable for one year when stored in a constant temperature freezer at −20 °C.